ENA387773021 Enamine

2-hydroxy-N-(2-methylphenyl)acetamide

  • MDL number MFCD08442914
  • Cas Number : 102879-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120593

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120593
Availability details for
Quantity
  • Availability
  • Add to Cart