ENA387773032 Enamine

5-methyl-1-(pyridin-4-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09702991
  • Cas Number : 241798-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120613

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120613
Availability details for
Quantity
  • Availability
  • Add to Cart