ENA387773033 Enamine

N-(3,5-dimethyl-1H-pyrazol-4-yl)-1-phenylmethanesulfonamide

  • MDL number MFCD12076576
  • Cas Number : 1187345-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120614

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120614
Availability details for
Quantity
  • Availability
  • Add to Cart