ENA387773035 Enamine

ethyl 2-{7-methylimidazo[1,2-a]pyrimidin-2-yl}acetate

  • MDL number MFCD11226453
  • Cas Number : 61830-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120616
Availability details for
Quantity
  • Availability
  • Add to Cart