ENA387773051 Enamine

methyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate

  • MDL number MFCD11111582
  • Cas Number : 1076197-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119089

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119089
Availability details for
Quantity
  • Availability
  • Add to Cart