ENA387773055 Enamine

[(5-cyclopropyl-1,3-oxazol-2-yl)methyl](methyl)amine

  • MDL number MFCD21130655
  • Cas Number : 1423034-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120675
Availability details for
Quantity
  • Availability
  • Add to Cart