ENA387773061 Enamine

1-(3-fluoropyridin-2-yl)piperazine

  • MDL number MFCD09910293
  • Cas Number : 85386-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120688

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P332+P313,P362,P363,P405,P501
hcodes C,Xn
MSDS https://www.enaminestore.com/catalog/EN300-120688
pictograms GHS05,GHS07
hstatements H302,H314,H315
Availability details for
Quantity
  • Availability
  • Add to Cart