ENA387773066 Enamine

1-[(2-nitrophenyl)sulfanyl]butan-2-one

  • MDL number MFCD16167319
  • Cas Number : 1249109-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120694

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120694
Availability details for
Quantity
  • Availability
  • Add to Cart