ENA387773069 Enamine

2-[(2,2,2-trifluoroethyl)sulfanyl]acetic acid

  • MDL number MFCD09737593
  • Cas Number : 675-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120700

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120700
Availability details for
Quantity
  • Availability
  • Add to Cart