ENA387773075 Enamine

1-methyl-1H-indole-3-sulfonamide

  • MDL number MFCD21884254
  • Cas Number : 1158209-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120707

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120707
Availability details for
Quantity
  • Availability
  • Add to Cart