ENA387773081 Enamine

4-hydroxy-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide

  • MDL number MFCD11164627
  • Cas Number : 515-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120724
Availability details for
Quantity
  • Availability
  • Add to Cart