ENA387773082 Enamine

1-(3,4-difluorophenyl)-2-methylpropan-1-amine

  • MDL number MFCD11148305
  • Cas Number : 1021031-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120725
Availability details for
Quantity
  • Availability
  • Add to Cart