ENA387773083 Enamine

1-(3,4-difluorophenyl)-2-methylpropan-1-one

  • MDL number MFCD11104170
  • Cas Number : 913719-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120726
Availability details for
Quantity
  • Availability
  • Add to Cart