ENA387773086 Enamine

3-amino-2-fluorophenol

  • MDL number MFCD16999409
  • Cas Number : 1186326-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112000
Availability details for
Quantity
  • Availability
  • Add to Cart