ENA387773089 Enamine

4-chloro-2-(4,5-dihydro-1,3-oxazol-2-yl)pyridine

  • MDL number MFCD22628379
  • Cas Number : 1423027-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119128

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-119128
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart