ENA387773092 Enamine

1-cyclopropyl-4-hydroxypyrrolidin-2-one

  • MDL number MFCD19665406
  • Cas Number : 1340512-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120753
Availability details for
Quantity
  • Availability
  • Add to Cart