ENA387773093 Enamine

4-cyclopropaneamido-2-methoxybenzene-1-sulfonyl chloride

  • MDL number MFCD09050011
  • Cas Number : 926187-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120766
Availability details for
Quantity
  • Availability
  • Add to Cart