ENA387773107 Enamine

1-[(3-chlorophenyl)methyl]-1H-pyrazol-5-amine

  • MDL number MFCD07366364
  • Cas Number : 956440-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57380
Availability details for
Quantity
  • Availability
  • Add to Cart