ENA387773108 Enamine

3-(3-aminopropyl)-1-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD22628560
  • Cas Number : 1170356-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57476
Availability details for
Quantity
  • Availability
  • Add to Cart