ENA387773122 Enamine

3,3-dimethylbutanenitrile

  • MDL number MFCD03413127
  • Cas Number : 3302-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74834

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-74834
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart