ENA387773133 Enamine

5-iodo-3-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD20543087
  • Cas Number : 289681-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85874
Availability details for
Quantity
  • Availability
  • Add to Cart