ENA387773144 Enamine

N-[(4-methylmorpholin-2-yl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD21643611
  • Cas Number : 1423035-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94044

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94044
Availability details for
Quantity
  • Availability
  • Add to Cart