ENA387773145 Enamine

5-(hydroxymethyl)-2-methylpyrimidin-4-ol

  • MDL number MFCD08704419
  • Cas Number : 698-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-119136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119136
Availability details for
Quantity
  • Availability
  • Add to Cart