ENA387773147 Enamine

1,2-oxazol-3-ol

  • MDL number MFCD16996230
  • Cas Number : 5777-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-100
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95046
Availability details for
Quantity
  • Availability
  • Add to Cart