ENA387773149 Enamine

3-(4-chlorophenyl)-2-methylprop-2-enoic acid

  • MDL number MFCD00136917
  • Cas Number : 1202-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95850

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95850
Availability details for
Quantity
  • Availability
  • Add to Cart