ENA387773152 Enamine

hexahydro-1H-pyrrolo[2,1-c][1,4]oxazine-3-carbonitrile

  • MDL number MFCD22628563
  • Cas Number : 1423025-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97079
Availability details for
Quantity
  • Availability
  • Add to Cart