ENA388191727 Enamine

2-isocyanato-2-methylpropanenitrile

  • MDL number MFCD22741186
  • Cas Number : 52161-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115685

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P270,P271,P280,P301+P310,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-115685
pictograms GHS02,GHS05,GHS06
hstatements H226,H301,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart