ENA388191728 Enamine

(1S)-2,2,2-trifluoro-1-(pyridin-3-yl)ethan-1-ol

  • MDL number MFCD22741189
  • Cas Number : 1226507-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116124
Availability details for
Quantity
  • Availability
  • Add to Cart