ENA388191730 Enamine

1-methyl-1-[2-(methylamino)ethyl]urea hydrochloride

  • MDL number MFCD22741192
  • Cas Number : 1423031-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116278

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116278
Availability details for
Quantity
  • Availability
  • Add to Cart