ENA388191739 Enamine

1-fluoro-3-methanesulfonyl-2-nitrobenzene

  • MDL number MFCD22741205
  • Cas Number : 1022971-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120330

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120330
Availability details for
Quantity
  • Availability
  • Add to Cart