ENA388191741 Enamine

(1S)-1-(1-benzothiophen-3-yl)-2,2,2-trifluoroethan-1-ol

  • MDL number MFCD22741207
  • Cas Number : 1423040-67-8
  • Molecular Weight : 232.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1S)-1-(1-benzothien-3-yl)-2,2,2-trifluoroethanol
Inchi Code 1S/C10H7F3OS/c11-10(12,13)9(14)7-5-15-8-4-2-1-3-6(7)8/h1-5,9,14H/t9-/m0/s1
Inchi Key WAYSUEKROXOKQO-VIFPVBQESA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-120544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120544
Availability details for
Quantity
  • Availability
  • Add to Cart