ENA388191745 Enamine

(4-chlorophenyl)(1,2,4-oxadiazol-3-yl)methanamine hydrochloride

  • MDL number MFCD22741212
  • Cas Number : 1423034-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120594

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120594
Availability details for
Quantity
  • Availability
  • Add to Cart