ENA388191753 Enamine

4-(difluoromethyl)-2-phenyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD22741222
  • Cas Number : 1423026-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120796
Availability details for
Quantity
  • Availability
  • Add to Cart