ENA388191756 Enamine

2-(2-phenyl-1,3-thiazol-4-yl)ethan-1-amine dihydrobromide

  • MDL number MFCD22741226
  • Cas Number : 250258-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120849

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120849
Availability details for
Quantity
  • Availability
  • Add to Cart