ENA388191768 Enamine

2-methyl-1-(2,2,2-trifluoroethyl)piperazine hydrochloride

  • MDL number MFCD22741243
  • Cas Number : 1423034-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121266
Availability details for
Quantity
  • Availability
  • Add to Cart