ENA388191777 Enamine

4-bromo-N-{[4-(trifluoromethyl)phenyl]methyl}aniline hydrochloride

  • MDL number MFCD22741258
  • Cas Number : 1423034-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121574
Availability details for
Quantity
  • Availability
  • Add to Cart