ENA388191780 Enamine

2-(tert-butoxy)-4-fluoro-1-nitrobenzene

  • MDL number MFCD22741263
  • Cas Number : 1369809-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121729

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121729
Availability details for
Quantity
  • Availability
  • Add to Cart