ENA388191785 Enamine

4-(carboxymethoxy)-1-phenyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD22741271
  • Cas Number : 1423026-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121852
Availability details for
Quantity
  • Availability
  • Add to Cart