ENA388191795 Enamine

4-(piperidin-2-yl)pyrrolidin-2-one

  • MDL number MFCD22741284
  • Cas Number : 1423024-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121943
Availability details for
Quantity
  • Availability
  • Add to Cart