ENA388191797 Enamine

methyl 2-[2-(hydroxyimino)cyclohexyl]acetate

  • MDL number MFCD22741286
  • Cas Number : 17448-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121980

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121980
Availability details for
Quantity
  • Availability
  • Add to Cart