ENA388191800 Enamine

1-(2-aminoethoxy)-4-bromo-2-(trifluoromethyl)benzene hydrochloride

  • MDL number MFCD22741290
  • Cas Number : 1423027-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122236

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122236
Availability details for
Quantity
  • Availability
  • Add to Cart