ENA388191804 Enamine

4-[4-(trifluoromethyl)phenyl]butan-1-amine hydrochloride

  • MDL number MFCD22741297
  • Cas Number : 1423034-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122308

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122308
Availability details for
Quantity
  • Availability
  • Add to Cart