ENA388191805 Enamine

ethyl 3-methyl-3-(methylamino)butanoate hydrochloride

  • MDL number MFCD22741301
  • Cas Number : 1423034-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122409
Availability details for
Quantity
  • Availability
  • Add to Cart