ENA388191817 Enamine

4-fluoro-3-methanesulfonyl-5-methylbenzoic acid

  • MDL number MFCD22741319
  • Cas Number : 1423033-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122894

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122894
Availability details for
Quantity
  • Availability
  • Add to Cart