ENA388191819 Enamine

5-(aminomethyl)-3-(3-methylbutyl)-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD22741321
  • Cas Number : 1423031-40-6
  • Molecular Weight : 222.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(aminomethyl)-3-isopentyl-1,3-oxazolidin-2-one hydrochloride
Inchi Code 1S/C9H18N2O2.ClH/c1-7(2)3-4-11-6-8(5-10)13-9(11)12;/h7-8H,3-6,10H2,1-2H3;1H
Inchi Key KXNPEOQHSNPNBD-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-122983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122983
Availability details for
Quantity
  • Availability
  • Add to Cart