ENA388191824 Enamine

tert-butyl N-[3-(hydrazinecarbonyl)propyl]carbamate

  • MDL number MFCD22741327
  • Cas Number : 860653-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66414
Availability details for
Quantity
  • Availability
  • Add to Cart