ENA388191830 Enamine

(1R)-7-(propan-2-yl)-1,2,3,4-tetrahydronaphthalen-1-ol

  • MDL number MFCD21297852
  • Cas Number : 1423040-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120534
Availability details for
Quantity
  • Availability
  • Add to Cart