ENA388191832 Enamine

(1S)-1-(6-methylpyridin-2-yl)ethan-1-amine

  • MDL number MFCD09256729
  • Cas Number : 1213399-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120545
Availability details for
Quantity
  • Availability
  • Add to Cart