ENA388191833 Enamine

2,2-dimethyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD09701428
  • Cas Number : 10489-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109248
Availability details for
Quantity
  • Availability
  • Add to Cart