ENA388191839 Enamine

(1S)-7-(propan-2-yl)-1,2,3,4-tetrahydronaphthalen-1-ol

  • MDL number MFCD21297853
  • Cas Number : 1423040-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120588

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120588
Availability details for
Quantity
  • Availability
  • Add to Cart